2-Bromo-6-methoxypyridine, 97%
2-Bromo-6-methoxypyridine, 97%
2-Bromo-6-methoxypyridine, 97%
Thermo Scientific Chemicals

2-Bromo-6-methoxypyridine, 97%

CAS: 40473-07-2 | C6H6BrNO | 188.02 g/mol
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 397670250
Price (JPY)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS40473-07-2
IUPAC Name2-bromo-6-methoxypyridine
Molecular FormulaC6H6BrNO
InChI KeyKMODISUYWZPVGV-UHFFFAOYSA-N
SMILESCOC1=CC=CC(Br)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to golden
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only