N-BOC-p-fenilenodiamina, 97 %, Thermo Scientific Chemicals
N-BOC-p-fenilenodiamina, 97 %, Thermo Scientific Chemicals
N-BOC-p-fenilenodiamina, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-p-fenilenodiamina, 97 %, Thermo Scientific Chemicals

CAS: 71026-66-9 | C11H16N2O2 | 208.26 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 398330250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS71026-66-9
IUPAC Nametert-butyl N-(4-aminophenyl)carbamate
Molecular FormulaC11H16N2O2
InChI KeyWIVYTYZCVWHWSH-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=CC=C(N)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Beige to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point113°C to 115°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only