N-BOC-D-Serine methyleester, 97%, Thermo Scientific Chemicals
N-BOC-D-Serine methyleester, 97%, Thermo Scientific Chemicals
N-BOC-D-Serine methyleester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-D-Serine methyleester, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
39937010010 gGlass Bottle
Catalog number 399370100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS95715-85-8
IUPAC Namemethyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoate
Molecular FormulaC9H17NO5
InChI KeySANNKFASHWONFD-UHFFFAOYNA-N
SMILESCOC(=O)C(CO)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Infrared spectrumConforms
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
Specific optical rotation+16° to +20° (20°C, 589 nm) (c=5% in Methanol)
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only