1-(2-Aminoethyl)piperazine, 99%
1-(2-Aminoethyl)piperazine, 99%
1-(2-Aminoethyl)piperazine, 99%
Thermo Scientific Chemicals

1-(2-Aminoethyl)piperazine, 99%

CAS: 140-31-8 | C6H15N3 | 129.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 400760025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS140-31-8
IUPAC Name2-(piperazin-1-yl)ethan-1-amine
Molecular FormulaC6H15N3
InChI KeyIMUDHTPIFIBORV-UHFFFAOYSA-N
SMILESNCCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4990 to 1.5010 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only