4-Aminophthalonitrile, 97%
4-Aminophthalonitrile, 97%
4-Aminophthalonitrile, 97%
Thermo Scientific Chemicals

4-Aminophthalonitrile, 97%

CAS: 56765-79-8 | C8H5N3 | 143.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Packaging:
Glass bottle
Catalog number 401020250
Price (USD)
618.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
618.00
Each
Chemical Identifiers
CAS56765-79-8
IUPAC Name4-aminobenzene-1,2-dicarbonitrile
Molecular FormulaC8H5N3
InChI KeyRRCAJFYQXKPXOJ-UHFFFAOYSA-N
SMILESNC1=CC=C(C#N)C(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
Appearance (Form)Crystalline powder or crystals and/or chunks
HPLC>=96.0 %
Infrared spectrumConforms
Melting point177°C to 182°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only