Benzoyl chloride, 98+%, ACS reagent
Benzoyl chloride, 98+%, ACS reagent
Benzoyl chloride, 98+%, ACS reagent
Thermo Scientific Chemicals

Benzoyl chloride, 98+%, ACS reagent

CAS: 98-88-4 | C7H5ClO | 140.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 402030050
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.50
Save 6.85
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
45.50
Save 6.85
Each
Chemical Identifiers
CAS98-88-4
IUPAC Namebenzoyl chloride
Molecular FormulaC7H5ClO
InChI KeyPASDCCFISLVPSO-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess NaOH98.0 to 100.5 %
Heavy metals (as Pb)=<10 ppm
Refractive index1.5520 to 1.5540 (20°C, 589 nm)
Residue after ignition=<0.005 %
Phosphorous compounds (as P)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only