1-Bromo-3-methylbutane, 99%
1-Bromo-3-methylbutane, 99%
1-Bromo-3-methylbutane, 99%
Thermo Scientific Chemicals

1-Bromo-3-methylbutane, 99%

CAS: 107-82-4 | C5H11Br | 151.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 403055000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-82-4
IUPAC Name1-bromo-3-methylbutane
Molecular FormulaC5H11Br
InChI KeyYXZFFTJAHVMMLF-UHFFFAOYSA-N
SMILESCC(C)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4390 to 1.4440 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only