Butyl formate, 97%
Butyl formate, 97%
Butyl formate, 97%
Thermo Scientific Chemicals

Butyl formate, 97%

CAS: 592-84-7 | C5H10O2 | 102.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
403541000100 gGlass bottle
Catalog number 403541000
Price (USD)
66.65
Special offer
Online exclusive
Ends: 28-Dec-2026
78.10
Save 11.45
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 28-Dec-2026
78.10
Save 11.45
Each
Chemical Identifiers
CAS592-84-7
IUPAC Namebutyl formate
Molecular FormulaC5H10O2
InChI KeyNMJJFJNHVMGPGM-UHFFFAOYSA-N
SMILESCCCCOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3880 to 1.3910 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only