Calcium hydroxide, 95+%, ACS reagent
Calcium hydroxide, 95+%, ACS reagent
Calcium hydroxide, 95+%, ACS reagent
Calcium hydroxide, 95+%, ACS reagent
Thermo Scientific Chemicals

Calcium hydroxide, 95+%, ACS reagent

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 403850025
Price (USD)
192.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
192.00
Each
Chemical Identifiers
CAS1305-62-0
IUPAC Namecalcium dihydroxide
Molecular FormulaCaH2O2
InChI KeyAXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (ICP-OES)=<30 ppm
Insoluble matter=<0.03 % (in HCl)
Sulfur compounds (as SO4)=<0.1 %
Iron (Fe)=<500 ppm
Chloride (Cl)=<300 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only