Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Thermo Scientific Chemicals

Cellulose Acetate Butyrate, Butyryl Content 35-39%

CAS: 9004-36-8 | (C12H14O10)n
Have Questions?
Catalog NumberQuantityPackaging
404255000500 gPlastic bottle
Catalog number 404255000
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Chemical Identifiers
CAS9004-36-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Viscosity17 to 28 s
Free acid (CH3COOH)=<0.03 %
Acetyl content12 to 15 %
Average molecular weight70000 (Mn)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only