Thermo Scientific Chemicals

hydrogen hexachloroplatinate(IV) hydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 405010250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

hydrogen hexachloroplatinate(IV) hydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 405010250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
26023-84-7
IUPAC Name
dihydrogen hexachloroplatinumtetrakis(ylium)
Molecular Formula
Cl6H2Pt
InChI Key
ZKOQTCCVSRSECD-UHFFFAOYSA-J
SMILES
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
Appearance (Form)
Powder and/or chunks
Solubility
(in alcohol) Passes test
Impurity
=<0.05 % Alkali and other salts (as sulfates)
Assay
>=37.50 % (Pt)
Suitability
(for K determination) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text