Copper(II) Trifluoromethanesulfonate, 98%
Copper(II) Trifluoromethanesulfonate, 98%
Copper(II) Trifluoromethanesulfonate, 98%
Thermo Scientific Chemicals

Copper(II) Trifluoromethanesulfonate, 98%

CAS: 34946-82-2 | C2CuF6O6S2 | 361.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 405420050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS34946-82-2
IUPAC Namecopper(II) triflate
Molecular FormulaC2H2CuF6O4S2
InChI KeyTXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES[Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey to light blue
Appearance (Form)Powder
Titration with Na2S2O397.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only