Corn oil
Thermo Scientific Chemicals

Corn oil

CAS: 8001-30-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 405435000
Price (USD)
54.65
Online exclusive
60.50
Save 5.85
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
54.65
Online exclusive
60.50
Save 5.85
Each
Chemical Identifiers
CAS8001-30-7
SpecificationsSpecification SheetSpecification Sheet
Acid value=<0.5 mg KOH/g
Appearance (Color)Clear yellow
Appearance (Form)Oily liquid
Infrared spectrumConforms
Refractive index1.4730 to 1.4760 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only