Coumarin 6, 98%, pure
Coumarin 6, 98%, pure
Coumarin 6, 98%, pure
Thermo Scientific Chemicals

Coumarin 6, 98%, pure

CAS: 38215-36-0 | C20H18N2O2S | 350.436 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Catalog number 405505000
Price (USD)
50.40
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
50.40
Each
Chemical Identifiers
CAS38215-36-0
IUPAC Name3-(1,3-benzothiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one
Molecular FormulaC20H18N2O2S
InChI KeyVBVAVBCYMYWNOU-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C2C=C(C3=NC4=CC=CC=C4S3)C(=O)OC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Infrared spectrumConforms
UV Lambda max455 to 461 nm (0.005g/l,Ethanol)
Appearance (Form)Powder
UV spectrophotometry>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only