Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent

CAS: 19004-19-4 | 2·5 H2O | 232.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 405855000
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS19004-19-4
IUPAC Namedicopper(2+) pentahydrate tetranitrate
Molecular FormulaCu2H10N4O17
InChI KeyIZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<50 ppm
Lead (Pb)=<10 ppm
Potassium (K)=<50 ppm
Appearance (Form)Crystalline powder
Insoluble matter=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only