Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution

CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol
数量:
250 g
1 kg
3 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 407292500
価格(JPY)
-
数量:
250 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS140-01-2
Color scale=<150 APHA
pH11.0 to 12.0 (1 % in water)
Appearance (Form)Clear solution
Titration39 to 41 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only