N,N-Dimethyl-p-phenylenediamine, 97%
N,N-Dimethyl-p-phenylenediamine, 97%
N,N-Dimethyl-p-phenylenediamine, 97%
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine, 97%

CAS: 99-98-9 | C8H12N2 | 136.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 408460250
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.70
Save 10.05
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
68.70
Save 10.05
Each
Chemical Identifiers
CAS99-98-9
IUPAC NameN1,N1-dimethylbenzene-1,4-diamine
Molecular FormulaC8H12N2
InChI KeyBZORFPDSXLZWJF-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Dark brown to black
GC>=96.0 %
Appearance (Form)Fused mass or liquid after melting
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only