trans,trans-Dibenzylideneacetone, 98%
trans,trans-Dibenzylideneacetone, 98%
trans,trans-Dibenzylideneacetone, 98%
Thermo Scientific Chemicals

trans,trans-Dibenzylideneacetone, 98%

CAS: 35225-79-7 | C17H14O | 234.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 409000250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35225-79-7
IUPAC Name(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
Molecular FormulaC17H14O
InChI KeyWMKGGPCROCCUDY-PHEQNACWSA-N
SMILESO=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point108°C to 114°C
Assay>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only