Ethylene dimethacrylate, 98%, stabilized
Ethylene dimethacrylate, 98%, stabilized
Ethylene dimethacrylate, 98%, stabilized
Thermo Scientific Chemicals

Ethylene dimethacrylate, 98%, stabilized

CAS: 97-90-5 | C10H14O4 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 409922500
Price (USD)
85.65
Online exclusive
94.50
Save 8.85
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
85.65
Online exclusive
94.50
Save 8.85
Each
Chemical Identifiers
CAS97-90-5
IUPAC Name2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate
Molecular FormulaC10H14O4
InChI KeySTVZJERGLQHEKB-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OCCOC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4530 to 1.4550 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only