Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Ethylenediaminetetraacetic acid, 99.5%, ACS reagent
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, 99.5%, ACS reagent

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 409930010
Price (USD)
152.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
152.00
Each
Chemical Identifiers
CAS60-00-4
IUPAC Name2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular FormulaC10H16N2O8
InChI KeyKCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILESOC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric99.4 to 100.6 %
Nitrilotriacetic acid=<0.1 % ((HOCOCH4)3N)
Iron (Fe)=<50 ppm
Appearance (Color)White
Insoluble matter=<0.005 % (in dilute NH4OH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only