FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 570, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 570, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 570, eBioscience™ - FAQs (41-5773-82, 41-5773-80)