Thermo Scientific Chemicals

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers, Thermo Scientific Chemicals

Catalog number: 410370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers, Thermo Scientific Chemicals

Catalog number: 410370250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
928-55-2
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
GC
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.3940 to 1.4000 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text