beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%
Thermo Scientific Chemicals

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%

beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8 | CAS: 13408-09-8 | C3H19Na2O11P | 308.13 g/mol
数量:
25 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 410990250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13408-09-8
IUPAC Name[(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
Molecular FormulaC3H19Na2O11P
InChI KeyVWVLGHQKEJESPW-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O
さらに表示
Appearance (Color)White
Infrared spectrumConforms
Isomer=<2 % alpha-Isomer
Appearance (Form)Crystalline powder
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only