Tetrachloroauric(III) acid trihydrate, ACS reagent
Tetrachloroauric(III) acid trihydrate, ACS reagent
Tetrachloroauric(III) acid trihydrate, ACS reagent
Thermo Scientific Chemicals

Tetrachloroauric(III) acid trihydrate, ACS reagent

Tetrachloroauric(III) Acid Trihydrate, >-49.0% (Au), HAuCl4.3H2O, CAS Number-16961-25-4, Gold trichloride Acid Trihydrate, Chloroauric Acid Trihydrate, 1g, Orange, 393.83, ACS Reagent, GHS Signal Word: Danger | CAS: 16961-25-4 | AuCl4H | 339.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number 411070250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16961-25-4
IUPAC Nametrichlorogold hydrochloride
Molecular FormulaAuCl4H
InChI KeyVDLSFRRYNGEBEJ-UHFFFAOYSA-K
SMILESCl.Cl[Au](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Appearance (Form)Powder and/or chunks
AppearanceMay have a brown hue
Alkalies+oth.metals (SO4)=<0.2 %
Insoluble matter=<0.1 % (10 % in ether)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only