Hydrazine acetate, 97%
Hydrazine acetate, 97%
Hydrazine acetate, 97%
Thermo Scientific Chemicals

Hydrazine acetate, 97%

CAS: 7335-65-1 | C2H8N2O2 | 92.10 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 411750250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS7335-65-1
IUPAC Nameacetic acid; hydrazine
Molecular FormulaC2H8N2O2
InChI KeyYFHNDHXQDJQEEE-UHFFFAOYSA-N
SMILESNN.CC(O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to yellow
Appearance (Form)Adhering powder or crystals
Infrared spectrumConforms
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only