Petroleum Ether, extra pure, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum Ether, extra pure, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum Ether, extra pure, boiling range 80-110°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Petroleum Ether, extra pure, boiling range 80-110°C, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantityPackaging
4131600202 kgEcoSafPak™
Catalog number 413160020
Price (TWD)
-
Quantity:
2 kg
Packaging:
EcoSafPak™
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0
IUPAC NamePetroleum ether
MDL NumberMFCD00081849
SpecificationsSpecification SheetSpecification Sheet
Additional infoBromine index < 100 ppm
Appearance (Form)Clear liquid
Color scale=<10 APHA
Refractive index1.3950 to 1.4030 (20°C, 589 nm)
Benzene=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only