Magnesium Chloride Hexahydrate, 99+%, ACS reagent
Magnesium Chloride Hexahydrate, 99+%, ACS reagent
Magnesium Chloride Hexahydrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Magnesium Chloride Hexahydrate, 99+%, ACS reagent

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 413415000
Price (EUR)
50,65
Online Exclusive
56,30
Save 5,65 (10%)
Each
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
50,65
Online Exclusive
56,30
Save 5,65 (10%)
Each
Chemical Identifiers
CAS7791-18-6
IUPAC Namemagnesium(2+) hexahydrate dichloride
Molecular FormulaCl2H12MgO6
InChI KeyDHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Sodium (Na)=<0.005 %
Ammonium (NH4)=<0.002 %
Manganese (Mn)=<5 ppm
Appearance (Color)White
Titration Complexometric99.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only