Methyl hexanoate, 99%, analytical standard for GC
Methyl hexanoate, 99%, analytical standard for GC
Methyl hexanoate, 99%, analytical standard for GC
Thermo Scientific Chemicals

Methyl hexanoate, 99%, analytical standard for GC

CAS: 106-70-7 | C7H14O2 | 130.19 g/mol
Have Questions?
Catalog NumberQuantity
4144200101 g
Catalog number 414420010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS106-70-7
IUPAC Namemethyl hexanoate
Molecular FormulaC7H14O2
InChI KeyNUKZAGXMHTUAFE-UHFFFAOYSA-N
SMILESCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only