Nile Red, 99%, pure
Nile Red, 99%, pure
Nile Red, 99%, pure
Thermo Scientific Chemicals

Nile Red, 99%, pure

CAS: 7385-67-3 | C20H18N2O2 | 318.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number 415710010
Price (USD)
575.65
Special offer
Online exclusive
Ends: 28-Dec-2026
677.00
Save 101.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
575.65
Special offer
Online exclusive
Ends: 28-Dec-2026
677.00
Save 101.35
Each
Chemical Identifiers
CAS7385-67-3
IUPAC Name8-(diethylamino)-12H-10-oxa-5-azatetraphen-12-one
Molecular FormulaC20H18N2O2
InChI KeyVOFUROIFQGPCGE-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark red to brown
Infrared spectrumConforms
UV spectrumComplies
UV spectrophotometry>=98.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only