4-Nitrobenzenesulfonyl chloride, 95%
4-Nitrobenzenesulfonyl chloride, 95%
4-Nitrobenzenesulfonyl chloride, 95%
Thermo Scientific Chemicals

4-Nitrobenzenesulfonyl chloride, 95%

CAS: 98-74-8 | C6H4ClNO4S | 221.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 415850250
Price (USD)
48.65
Special offer
Online exclusive
Ends: 28-Dec-2026
57.50
Save 8.85
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 28-Dec-2026
57.50
Save 8.85
Each
Chemical Identifiers
CAS98-74-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point75°C to 80°C
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only