Pararosaniline chloride, 96%, pure
Pararosaniline chloride, 96%, pure
Pararosaniline chloride, 96%, pure
Thermo Scientific Chemicals

Pararosaniline chloride, 96%, pure

CAS: 569-61-9 | C19H17N3·HCl | 323.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 416801000
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Chemical Identifiers
CAS569-61-9
IUPAC Namehydrogen 4-[(4-aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular FormulaC19H18ClN3
InChI KeyJUQPZRLQQYSMEQ-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(C=C1)C(C1=CC=C(N)C=C1)=C1C=CC(=N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV Lambda max543 to 549 nm (c=0.0003%, 50% EtOH)
HPLC>=95 %
Appearance (Color)Green to dark green
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS.
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only