Thermo Scientific Chemicals

Quinine sulfate dihydrate, specified according to the requirements of USP, Thermo Scientific Chemicals

Catalog number: 418781000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Quinine sulfate dihydrate, specified according to the requirements of USP, Thermo Scientific Chemicals

Catalog number: 418781000
100 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
6119-70-6
IUPAC Name
bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Molecular Formula
C40H54N4O10S
InChI Key
ZHNFLHYOFXQIOW-OIGVVMIYNA-N
SMILES
O.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
Appearance (Color)
White
Identification B
Passes test
Titration with HClO4
99.0 to 101.0 % (on anhydrous basis)
Residue after ignition
=<0.1 %
Water
4.0 to 5.5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text