Rosin, gum
Rosin, gum
Rosin, gum
Thermo Scientific Chemicals

Rosin, gum

CAS: 8050-09-7 | C20H30O2 | 302.46 g/mol
数量:
500 g
2.5 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 419175000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS8050-09-7
IUPAC Name1,4a-dimethyl-7-(propan-2-yl)-1,2,3,4,4a,4b,5,6,10,10a-decahydrophenanthrene-1-carboxylic acid
Molecular FormulaC20H30O2
InChI KeyRSWGJHLUYNHPMX-UHFFFAOYNA-N
SMILESCC(C)C1=CC2=CCC3C(C)(CCCC3(C)C(O)=O)C2CC1
さらに表示
Appearance (Color)Almost white to yellow to orange
Appearance (Form)Powder or flakes or chunks
Infrared spectrumConforms
Acid value156 to 182 mg KOH/g
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only