CD4 Monoclonal Antibody (RM4-5), eFluor™ 615, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RM4-5), eFluor™ 615, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RM4-5), eFluor™ 615, eBioscience™ - FAQs (42-0042-80, 42-0042-82)