alpha Tubulin Monoclonal Antibody (DM1A), eFluor™ 615, eBioscience™ - FAQs
alpha Tubulin Monoclonal Antibody (DM1A), eFluor™ 615, eBioscience™ - FAQs
View additional product information for alpha Tubulin Monoclonal Antibody (DM1A), eFluor™ 615, eBioscience™ - FAQs (42-4502-82)