Tetrabutylammonium Hydroxide Titrant, 0.4M in Water, HPLC Grade
Tetrabutylammonium Hydroxide Titrant, 0.4M in Water, HPLC Grade
Tetrabutylammonium Hydroxide Titrant, 0.4M in Water, HPLC Grade
Thermo Scientific Chemicals

Tetrabutylammonium Hydroxide Titrant, 0.4M in Water, HPLC Grade

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 420120025
Price (USD)
1,251.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,472.00
Save 220.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
1,251.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,472.00
Save 220.35
Each
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 420 nm A: =<0.15
Color scale=<10 APHA
Bromide (Br)=<500 ppm
Appearance (Form)Clear solution
Titration with HCl0.37 to 0.43 M
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalyst
RUO – Research Use Only