Tetraethylammonium nitrate, 98%
Tetraethylammonium nitrate, 98%
Tetraethylammonium nitrate, 98%
Thermo Scientific Chemicals

Tetraethylammonium nitrate, 98%

CAS: 1941-26-0 | C8H20N2O3 | 192.26 g/mol
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 420310250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1941-26-0
IUPAC Nametetraethylazanium nitrate
Molecular FormulaC8H20N2O3
InChI KeyJTJKNAJRGLQKDZ-UHFFFAOYSA-N
SMILES[O-][N+]([O-])=O.CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellowish-white
Appearance (Form)Crystalline powder
Titration with HClO497.5 to 102.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only