p-Toluenesulfonic acid, pure, 12% in acetic acid
p-Toluenesulfonic acid, pure, 12% in acetic acid
p-Toluenesulfonic acid, pure, 12% in acetic acid
Acros Organics

p-Toluenesulfonic acid, pure, 12% in acetic acid

CAS: 104-15-4 | C7H8O3S | 172.198 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 421220010
also known as 42122
HSN 29041090
Price (INR)
36,813.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (INR)
36,813.00
Each
Chemical Identifiers
CAS104-15-4
IUPAC Name4-methylbenzene-1-sulfonic acid
Molecular FormulaC7H8O3S
InChI KeyJOXIMZWYDAKGHI-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow to pink
Titration with NaOH11 to 13 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only