Trichloroethylene, 99.6%, ACS reagent, stabilized
Trichloroethylene, 99.6%, ACS reagent, stabilized
Trichloroethylene, 99.6%, ACS reagent, stabilized
Trichloroethylene, 99.6%, ACS reagent, stabilized
Thermo Scientific Chemicals

Trichloroethylene, 99.6%, ACS reagent, stabilized

CAS: 79-01-6 | C2HCl3 | 131.39 g/mol
数量:
500 mL
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 421525000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS79-01-6
IUPAC Name1,1,2-trichloroethene
Molecular FormulaC2HCl3
InChI KeyXSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILESClC=C(Cl)Cl
さらに表示
Color scale=<10 APHA
Water=<0.02 % (Coulometric)
Titrable acid=<0.0001 meq/g
Free halogensPasses test
Heavy metals (as Pb)=<1 ppm
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only