N,2,3-Trimethyl-2-isopropylbutamide
N,2,3-Trimethyl-2-isopropylbutamide
N,2,3-Trimethyl-2-isopropylbutamide
Thermo Scientific Chemicals

N,2,3-Trimethyl-2-isopropylbutamide

CAS: 51115-67-4 | C10H21NO | 171.28 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 421930050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS51115-67-4
IUPAC NameN,2,3-trimethyl-2-(propan-2-yl)butanamide
Molecular FormulaC10H21NO
InChI KeyRWAXQWRDVUOOGG-UHFFFAOYSA-N
SMILESCNC(=O)C(C)(C(C)C)C(C)C
さらに表示
Infrared spectrumConforms
GC>=99.4 %
Appearance (Color)White
Water=<0.5 %
Appearance (Form)Powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only