Calcium oxide, 97+%, for analysis, powder
Calcium oxide, 97+%, for analysis, powder
Calcium oxide, 97+%, for analysis, powder
Thermo Scientific Chemicals

Calcium oxide, 97+%, for analysis, powder

Calcium oxide, 97+%, CaO, CAS Number-1305-78-8, gebrannter kalk, calxyl, calcia, burnt lime, calcium oxide, airlock, calcium monoxide, calx, unslaked lime, 1kg, 2.0% max., 97% min. (CaO) on dry substance (Complexometry), 2850 deg.C, CHEBI:31344, White or Yellow, 215-138-9, 56.08, Analysis | CAS: 1305-78-8 | CaO | 56.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 422830025
Price (USD)
66.10
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
66.10
Each
Chemical Identifiers
CAS1305-78-8
IUPAC Namecalcium oxidandiide
Molecular FormulaCaO
InChI KeyBRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES[O--].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric>=97.0 % (CaO) on dry substance
Heavy metals (as Pb)=<100 ppm
Loss on ignition=<5 %
NH4OH precipitate=<2.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only