Copper(II) sulfate pentahydrate, 98+%, ACS reagent
Copper(II) sulfate pentahydrate, 98+%, ACS reagent
Copper(II) sulfate pentahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Copper(II) sulfate pentahydrate, 98+%, ACS reagent

CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
3 kg
Packaging:
Plastic bottle
Catalog number 423615000
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Chemical Identifiers
CAS7758-99-8
IUPAC Namecopper(2+) pentahydrate sulfate
Molecular FormulaCuH10O9S
InChI KeyJZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
Nitrogen compounds (as N)=<0.002 %
Chloride (Cl)=<10 ppm
Iron (Fe)=<30 ppm
Calcium (Ca)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only