Mercury(II) chloride, 99.5+%, ACS reagent
Mercury(II) chloride, 99.5+%, ACS reagent
Mercury(II) chloride, 99.5+%, ACS reagent
Thermo Scientific Chemicals

Mercury(II) chloride, 99.5+%, ACS reagent

CAS: 7487-94-7 | Cl2Hg | 271.49 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 423931000
価格(JPY)
-
数量:
100 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7487-94-7
IUPAC Namemercury(2+) dichloride
Molecular FormulaCl2Hg
InChI KeyLWJROJCJINYWOX-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hg++]
さらに表示
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration Complexometric>=99.5 %
Residue after reduction=<0.02 %
Iron (Fe)=<20 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only