Potassium thiocyanate, 99%, ACS reagent
Potassium thiocyanate, 99%, ACS reagent
Potassium thiocyanate, 99%, ACS reagent
Thermo Scientific Chemicals

Potassium thiocyanate, 99%, ACS reagent

CAS: 333-20-0 | CKNS | 97.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 424235000
Price (USD)
126.65
Online Exclusive
141.00
Save 14.35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
141.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS333-20-0
IUPAC Namepotassium cyanosulfanide
Molecular FormulaCKNS
InChI KeyZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES[K+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only