Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Thermo Scientific Chemicals

Toluene, 99.5%, ACS reagent, meets the requirements of Reag. Ph.Eur.

CAS: 108-88-3 | C7H8 | 92.14 g/mol
数量:
500 mL
1 L
2.5 L
25 L
パッケージ:
すべて表示
EcoSafPak™ Glass Bottle
製品番号(カタログ番号) 424555000
価格(JPY)
-
数量:
500 mL
パッケージ:
EcoSafPak™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
さらに表示
Color scale=<10 APHA
Density(20/20°C) 0.865 to 0.870
Boiling point109°C to 111°C
Residue after evaporation=<0.001 %
Sulfur compounds (as S)=<0.003 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only