3-Methylsulphonylaniline hydrochloride, 95%
3-Methylsulphonylaniline hydrochloride, 95%
3-Methylsulphonylaniline hydrochloride, 95%
Thermo Scientific Chemicals

3-Methylsulphonylaniline hydrochloride, 95%

CAS: 80213-28-1 | C7H10ClNO2S | 207.67 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4248700252.5 gGlass bottle
Catalog number 424870025
Price (USD)
-
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS80213-28-1
IUPAC Namehydrogen 3-methanesulfonylaniline chloride
Molecular FormulaC7H10ClNO2S
InChI KeyZMPXGSYLKFAGOV-UHFFFAOYSA-N
SMILES[H+].[Cl-].CS(=O)(=O)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Off-white to light beige
Appearance (Form)Powder
Titration Argentometric>=94 %
Loss on drying=<5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only