Ethyl isonicotinoylacetate, 95%
Ethyl isonicotinoylacetate, 95%
Ethyl isonicotinoylacetate, 95%
Thermo Scientific Chemicals

Ethyl isonicotinoylacetate, 95%

CAS: 26377-17-3 | C10H11NO3 | 193.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 425480025
Price (USD)
86.70
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
86.70
Each
Chemical Identifiers
CAS26377-17-3
IUPAC Nameethyl 3-oxo-3-(pyridin-4-yl)propanoate
Molecular FormulaC10H11NO3
InChI KeyPCJNYGPKMQQCPX-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point53°C to 59°C
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only