Neopentyl bromide, 98%
Neopentyl bromide, 98%
Neopentyl bromide, 98%
Thermo Scientific Chemicals

Neopentyl bromide, 98%

CAS: 630-17-1 | C5H11Br | 151.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 425720250
Price (USD)
220.65
Special offer
Online exclusive
Ends: 28-Dec-2026
259.00
Save 38.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
220.65
Special offer
Online exclusive
Ends: 28-Dec-2026
259.00
Save 38.35
Each
Chemical Identifiers
CAS630-17-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4350 to 1.4390 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only