Dimethyl chlorophosphate, 96%
Dimethyl chlorophosphate, 96%
Dimethyl chlorophosphate, 96%
Thermo Scientific Chemicals

Dimethyl chlorophosphate, 96%

CAS: 813-77-4 | C2H6ClO3P | 144.49 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 426140050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS813-77-4
IUPAC Namedimethyl phosphorochloridate
Molecular FormulaC2H6ClO3P
InChI KeyNGFFLHMFSINFGB-UHFFFAOYSA-N
SMILESCOP(Cl)(=O)OC
檢視更多
規格Specification Sheet規格表
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
Titration Argentometric>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only