1,3,2-Dioxathiolane 2,2-dioxide, 96%, Thermo Scientific Chemicals
1,3,2-Dioxathiolane 2,2-dioxide, 96%, Thermo Scientific Chemicals
1,3,2-Dioxathiolane 2,2-dioxide, 96%, Thermo Scientific Chemicals
1,3,2-Dioxathiolane 2,2-dioxide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,2-Dioxathiolane 2,2-dioxide, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
4266300505 gGlass bottle
Catalog number 426630050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1072-53-3
IUPAC Name1,3,2λ⁶-dioxathiolane-2,2-dione
Molecular FormulaC2H4O4S
InChI KeyZPFAVCIQZKRBGF-UHFFFAOYSA-N
SMILESO=S1(=O)OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Melting point95°C to 99°C
Infrared spectrumConforms
Appearance (Form)Powder
GC>=95.0 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only